Search
Search

Hoxb9| Gene Symbol: Hoxb9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008270.2 | NP_032296.2 | 2 | 886 | ||
| GenBank mRNA | AI116698.1 | - | 2 | 267 | ||
| AK078860.1 | - | 1 | 285 | |||
| BC100742.1 | - | 2 | 239 | |||
| BC100743.1 | - | 2 | 454 | 40045678 | ||
| BC100744.1 | - | 2 | 239 | |||
| BC103573.1 | - | 2 | 454 | 40045679 | ||
| M34857.1 | - | 2 | 305 | |||
| S66855.1 | - | 2 | 707 | |||