Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hsd17b4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008292.4 | NP_032318.2 | 22 | 2072 | ||
GenBank mRNA | AK004866.1 | - | 22 | 2072 | ||
AK088381.1 | - | 22 | 2047 | |||
AK147182.1 | - | 22 | 2044 | |||
AK152532.1 | - | 22 | 1966 | |||
AK161655.1 | - | 22 | 1967 | |||
AK166801.1 | - | 22 | 1985 | |||
AK167060.1 | - | 22 | 2022 | |||
AK169077.1 | - | 22 | 1974 | |||
BC022175.1 | - | 22 | 1945 | 4209394 | ||
X89998.1 | - | 22 | 1933 |