Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hspd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010477.4 | NP_034607.3 | 6 | 781 | ||
GenBank mRNA | AK088844.1 | - | 6 | 695 | ||
AK143882.1 | - | 6 | 686 | |||
AK146831.1 | - | 6 | 733 | |||
AK177219.1 | - | 3 | 206 | |||
AK184625.1 | - | 3 | 212 | |||
AK187612.1 | - | 3 | 211 | |||
AK188312.1 | - | 3 | 205 | |||
AK216069.1 | - | 3 | 175 | |||
BC016400.1 | - | 6 | 667 | 3585270 | ||
BC106112.1 | - | 6 | 682 | |||
X53584.1 | - | 5 | 571 | |||
X55023.1 | - | 6 | 643 |