Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other HSP90AA1 SIRNA Assays ›
          Gene Symbol:
          Hsp90aa1
          Gene Name:
          heat shock protein 90, alpha (cytosolic), class A member 1
          Gene Aliases:
          86kDa, 89kDa, AL024080, AL024147, Hsp86-1, Hsp89, Hsp90, Hspca, hsp4
          Chromosome Location:
          Chr.12: 110691036 - 110696395 on Build GRCm38
          UniGene ID:
          Mm.1843
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          siRNA ID s67900

          Purification
          Size
          Availability Made To Order
          Catalog # 4390771
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Hsp90aa1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_010480.5 NP_034610.1 8 1536
          GenBank mRNA AK004658.1 - 8 1538
          AK077095.1 - 8 1459
          AK146944.1 - 8 1451
          AK147641.1 - 8 1410
          AK157849.1 - 8 1452
          AK158753.1 - 7 1486
          AK161276.1 - 8 1481
          AK166696.1 - 8 1451
          AK166720.1 - 8 1451
          AK166729.1 - 8 1442
          AK166847.1 - 8 1441
          AK166867.1 - 8 1450
          AK166896.1 - 8 1455
          AK167008.1 - 8 1451
          AK167065.1 - 8 1451
          AK167086.1 - 8 1450
          AK167114.1 - 8 1451
          AK167165.1 - 5 861
          AK167171.1 - 8 1450
          AK167188.1 - 8 1451
          AK167293.1 - 5 861
          AK168407.1 - 8 1451
          AK168605.1 - 8 1454
          AK168620.1 - 8 1451
          BC018232.1 - 8 1416
          BC046614.1 - 8 1418 6491307
          BC049124.1 - 8 1531 6816250
          BC057287.1 - 8 1434
          BC090391.1 - 4 464
          BC094024.1 - 8 1433 4501952
          X16857.1 - 1 16

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          Hsp90 family chaperone

          Gene Ontology Categories:

          Function(s) Process(es)

          neuron migration
          protein folding
          nitric oxide biosynthetic process
          response to stress
          response to unfolded protein
          positive regulation of lamellipodium assembly
          positive regulation of protein import into nucleus, translocation
          protein refolding
          protein import into mitochondrial outer membrane
          positive regulation of nitric oxide biosynthetic process
          positive regulation of cytotoxic T cell differentiation
          positive regulation of cell size
          regulation of catalytic activity
          chaperone-mediated protein complex assembly
          nucleotide binding
          UTP binding
          CTP binding
          mRNA binding
          protein binding
          ATP binding
          GTP binding
          ATPase activity
          sulfonylurea receptor binding
          protein kinase binding
          protein phosphatase binding
          MHC class II protein complex binding
          nitric-oxide synthase regulator activity
          TPR domain binding
          dATP binding
          identical protein binding
          protein homodimerization activity
          ion channel binding
          poly(A) RNA binding
          GTPase binding
          Rho GDP-dissociation inhibitor binding
          unfolded protein binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-gwzxr:80/100.66.76.73:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0