Search
Search

Idh1| Gene Symbol: Idh1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001111320.1 | NP_001104790.1 | 8 | 1215 | ||
| NM_010497.3 | NP_034627.3 | 8 | 1193 | |||
| GenBank mRNA | AF020039.1 | - | 7 | 1043 | ||
| AK087063.1 | - | 8 | 1095 | |||
| AK149019.1 | - | 8 | 1124 | |||
| AK151212.1 | - | 8 | 1118 | |||
| AK159173.1 | - | 8 | 1124 | |||
| AK160896.1 | - | 8 | 1125 | |||
| AK167158.1 | - | 8 | 1165 | |||
| AK167583.1 | - | 8 | 1129 | |||
| AK168832.1 | - | 8 | 1166 | |||
| AK216686.1 | - | 2 | 49 | |||
| BC088986.1 | - | 8 | 1165 | 6808731 | ||
| DQ403075.1 | - | 7 | 943 | |||