Search
Search

HOXB9| Gene Symbol: HOXB9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024017.4 | NP_076922.1 | 1 | 648 | ||
| GenBank mRNA | AK056123.1 | - | 1 | 427 | ||
| AK056414.1 | - | 2 | 471 | |||
| AK315480.1 | - | 1 | 506 | |||
| BC015565.1 | - | 1 | 539 | 4139164 | ||
| BF305592.1 | - | 1 | 529 | |||