Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Igh-VS107 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC111444.1 | - | 1 | 122 | ||
M16334.1 | - | 1 | 51 | |||
M37679.1 | - | 1 | 41 |