Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other IL6 SIRNA Assays ›
          Gene Symbol:
          Il6
          Gene Name:
          interleukin 6
          Gene Aliases:
          Il-6
          Chromosome Location:
          Chr.5: 30013161 - 30019968 on Build GRCm38
          UniGene ID:
          Mm.1019
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          siRNA ID s68292

          Purification
          Size
          Availability Made To Order
          Catalog # 4390771
          Price 5,354.00
          Your Price
          Online offer:
          5354.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Il6
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_031168.1 NP_112445.1 3 336
          GenBank mRNA AK089780.1 - 3 355
          AK150440.1 - 3 355
          AK152189.1 - 2 342
          BC132458.1 - 3 329 40130735
          BC138766.1 - 3 329 8861788
          BC145409.1 - 3 329
          DQ788722.1 - 3 305
          J03783.1 - 3 353
          X06203.1 - 3 338
          X54542.1 - 3 336

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          neutrophil apoptotic process
          myeloid cell homeostasis
          hepatic immune response
          positive regulation of acute inflammatory response
          negative regulation of protein kinase activity
          acute-phase response
          inflammatory response
          immune response
          positive regulation of cell proliferation
          negative regulation of cell proliferation
          regulation of cell shape
          response to wounding
          regulation of vascular endothelial growth factor production
          positive regulation of gene expression
          cell growth
          cytokine-mediated signaling pathway
          endocrine pancreas development
          neuron projection development
          positive regulation of chemokine production
          positive regulation of interleukin-6 production
          negative regulation of collagen biosynthetic process
          positive regulation of peptidyl-serine phosphorylation
          positive regulation of protein import into nucleus, translocation
          positive regulation of T cell proliferation
          T cell activation
          regulation of cell proliferation
          positive regulation of tyrosine phosphorylation of Stat3 protein
          glucose homeostasis
          defense response to protozoan
          regulation of apoptotic process
          positive regulation of apoptotic process
          negative regulation of apoptotic process
          negative regulation of cysteine-type endopeptidase activity involved in apoptotic process
          positive regulation of MAPK cascade
          negative regulation of chemokine biosynthetic process
          regulation of circadian sleep/wake cycle, non-REM sleep
          positive regulation of nitric oxide biosynthetic process
          cell redox homeostasis
          positive regulation of T-helper 2 cell differentiation
          positive regulation of neuron differentiation
          negative regulation of gluconeogenesis
          positive regulation of translation
          positive regulation of DNA replication
          positive regulation of transcription, DNA-templated
          positive regulation of transcription from RNA polymerase II promoter
          positive regulation of JAK-STAT cascade
          muscle cell cellular homeostasis
          negative regulation of hormone secretion
          negative regulation of muscle organ development
          positive regulation of smooth muscle cell proliferation
          positive regulation of epithelial cell proliferation
          negative regulation of cytokine secretion
          positive regulation of peptidyl-tyrosine phosphorylation
          positive regulation of B cell activation
          positive regulation of immunoglobulin secretion
          positive regulation of sequence-specific DNA binding transcription factor activity
          response to glucocorticoid
          defense response to virus
          positive regulation of protein kinase B signaling
          positive regulation of transmission of nerve impulse
          branching involved in salivary gland morphogenesis
          epithelial cell proliferation involved in salivary gland morphogenesis
          glucagon secretion
          interleukin-6-mediated signaling pathway
          cellular response to hydrogen peroxide
          positive regulation of ERK1 and ERK2 cascade
          cellular response to lipopolysaccharide
          cellular response to interleukin-1
          cellular response to tumor necrosis factor
          T-helper 17 cell lineage commitment
          negative regulation of neuron death
          receptor binding
          cytokine activity
          interleukin-6 receptor binding
          protein binding
          growth factor activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-2k58s:80/100.66.76.192:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0