Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Inpp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008384.2 | NP_032410.2 | 3 | 585 | ||
GenBank mRNA | AK078382.1 | - | 3 | 473 | ||
BC025072.1 | - | 3 | 330 | 4456812 | ||
U27295.1 | - | 3 | 434 |