Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other ITGB1BP1 SIRNA Assays ›
          Gene Symbol:
          Itgb1bp1
          Gene Name:
          integrin beta 1 binding protein 1
          Gene Aliases:
          AI449260, AU019480, Icap1
          Chromosome Location:
          Chr.12: 21267246 - 21286279 on Build GRCm38
          UniGene ID:
          Mm.273333
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          siRNA ID s68457

          Purification
          Size
          Availability Made To Order
          Catalog # 4390771
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Itgb1bp1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_008403.4 NP_032429.1 2 241
          GenBank mRNA AJ001373.1 - 2 223
          AK002786.1 - 2 198
          AK004172.1 - 2 197
          AK007635.1 - 2 199
          AK075577.1 - 2 242
          AK089937.1 - 2 355
          AK147929.1 - 2 190
          BC021151.1 - 2 380
          BC028772.1 - 1 77 1230640

          Back To Top

          More Information


          Other Species Targeted

          This siRNA also targets the following transcripts in Rat: NM_001106719, XM_006239979, XM_006239981, XM_006239980, BC169104, and others.

          Gene Ontology Categories:

          Function(s) Process(es)

          angiogenesis
          blood vessel endothelial cell proliferation involved in sprouting angiogenesis
          transcription, DNA-templated
          regulation of transcription, DNA-templated
          negative regulation of protein kinase activity
          negative regulation of cell adhesion involved in substrate-bound cell migration
          cell adhesion
          cell-matrix adhesion
          Notch signaling pathway
          integrin-mediated signaling pathway
          positive regulation of cell proliferation
          negative regulation of cell proliferation
          positive regulation of endothelial cell migration
          negative regulation of fibroblast migration
          protein transport
          cell differentiation
          regulation of cell adhesion
          biomineral tissue development
          negative regulation of protein binding
          activation of protein kinase B activity
          integrin activation
          regulation of cell adhesion mediated by integrin
          tube formation
          cellular response to vascular endothelial growth factor stimulus
          regulation of GTPase activity
          receptor clustering
          cellular response to fibroblast growth factor stimulus
          positive regulation of Notch signaling pathway
          positive regulation of transcription from RNA polymerase II promoter
          regulation of blood vessel size
          myoblast migration
          positive regulation of stress fiber assembly
          positive regulation of cell division
          positive regulation of focal adhesion assembly
          negative regulation of focal adhesion assembly
          positive regulation of protein kinase B signaling
          negative regulation of ERK1 and ERK2 cascade
          negative regulation of cell migration involved in sprouting angiogenesis
          positive regulation of protein targeting to membrane
          negative regulation of protein targeting to membrane
          negative regulation of substrate adhesion-dependent cell spreading
          regulation of integrin-mediated signaling pathway
          GDP-dissociation inhibitor activity
          integrin binding
          protein binding
          protein C-terminus binding
          protein transporter activity
          kinase binding
          protein kinase binding
          protein complex binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-tlt54:80/100.66.79.31:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0