Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Itpr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010585.5 | NP_034715.3 | 3, 4 | 444 | ||
GenBank mRNA | AK042240.1 | - | 3, 4 | 299 | ||
AK047719.1 | - | 3, 4 | 445 | |||
AK137477.1 | - | 3, 4 | 446 | |||
X15373.1 | - | 3, 4 | 422 |