Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kcnk2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001159850.1 | NP_001153322.1 | 7 | 1484 | ||
NM_001281847.1 | NP_001268776.1 | 7 | 1375 | |||
NM_001281848.1 | NP_001268777.1 | 6 | 1216 | |||
NM_010607.3 | NP_034737.2 | 7 | 1580 | |||
GenBank mRNA | AK138427.1 | - | 7 | 1256 | ||
AK140716.1 | - | 7 | 1255 | |||
AK164197.1 | - | 6 | 1216 | |||
AY736359.1 | - | 7 | 1219 | |||
BC062094.1 | - | 7 | 1198 | 6848897 | ||
BQ572833.1 | - | 1 | 56 | |||
CN528767.1 | - | 4 | 601 | |||
U73488.2 | - | 8 | 1657 |