Search Thermo Fisher Scientific
Gene Symbol: Klra2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001170851.1 | NP_001164322.1 | 5 | 696 | ||
NM_008462.5 | NP_032488.4 | 5 | 696 | |||
GenBank mRNA | AF395446.1 | - | 4 | 530 | ||
AK017140.1 | - | 5 | 687 | |||
BC064711.1 | - | 6 | 696 | 30295755 | ||
EF025059.1 | - | 5 | 530 |