Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Klrd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010654.3 | NP_034784.1 | 6 | 452 | ||
GenBank mRNA | AF030311.1 | - | 5 | 432 | ||
AF039025.1 | - | 6 | 446 | |||
AF057714.1 | - | 5 | 432 | |||
AK136548.1 | - | 6 | 453 | |||
AK162976.1 | - | 7 | 513 | |||
BC117112.1 | - | 6 | 470 | 40126431 | ||
BC120854.1 | - | 6 | 472 | 40129777 |