Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sspn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010656.2 | NP_034786.1 | 1 | 215 | ||
GenBank mRNA | AK148179.1 | - | 1 | 217 | ||
AK162864.1 | - | 1 | 240 | |||
AK163041.1 | - | 1 | 241 | |||
AK164430.1 | - | 1 | 241 | |||
AK164529.1 | - | 1 | 187 | |||
BC021484.1 | - | 1 | 240 | 5344514 | ||
U02487.1 | - | 1 | 215 |