Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Krtap6-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010672.3 | NP_034802.1 | 1 | 392 | ||
GenBank mRNA | BC107313.1 | - | 1 | 392 | 40053340 | |
BC107314.1 | - | 1 | 392 | 40053341 | ||
BC114404.1 | - | 1 | 390 | |||
BC114532.1 | - | 1 | 390 | |||
BC127045.1 | - | 1 | 392 | |||
BC127046.1 | - | 1 | 392 | |||
CJ048388.1 | - | 1 | 432 | |||
D86420.1 | - | 1 | 392 |