siRNA Details
Gene Symbol: L1cam | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008478.3 | NP_032504.3 | 6, 7 | 779 | ||
GenBank mRNA | AK139407.1 | - | 6, 7 | 741 | ||
BC056988.1 | - | 6, 7 | 704 | 6822217 |

More Information
Other Species Targeted

Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Set Membership: |
![]() |
Panther Classification:
