Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lama1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008480.2 | NP_032506.2 | 45 | 6521 | ||
GenBank mRNA | CN461817.1 | - | 6 | 651 | ||
J04064.1 | - | 46 | 6502 |