Search
Search

Lipe| Gene Symbol: Lipe | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001039507.2 | NP_001034596.1 | 2 | 538 | ||
| NM_010719.5 | NP_034849.2 | 2 | 974 | |||
| GenBank mRNA | AK019846.1 | - | 2 | 1375 | ||
| AK029984.1 | - | 2 | 977 | |||
| AK169858.1 | - | 2 | 533 | |||
| BC021642.1 | - | 2 | 534 | 4188567 | ||
| BC053502.1 | - | 2 | 598 | |||
| U08188.1 | - | 1 | 1059 | |||