Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other LPL SIRNA Assays ›
          Gene Symbol:
          Lpl
          Gene Name:
          lipoprotein lipase
          Gene Aliases:
          -
          Chromosome Location:
          Chr.8: 68880555 - 68906932 on Build GRCm38
          UniGene ID:
          Mm.1514
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          siRNA ID s69302

          Purification
          Size
          Availability Made To Order
          Catalog # 4390771
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Lpl
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_008509.2 NP_032535.2 8 1349
          GenBank mRNA AK002645.1 - 8 1275
          AK017272.1 - 1347
          AK045064.1 - 8 1238
          AK079446.1 - 8 1238
          AK086023.1 - 8 1170
          AK150328.1 - 8 1350
          AK150355.1 - 8 1350
          AK150375.1 - 8 1348
          AK150457.1 - 8 1245
          AK150488.1 - 8 1350
          AK150505.1 - 8 1350
          AK150577.1 - 8 1350
          AK150585.1 - 8 1350
          AK150593.1 - 8 1350
          AK150646.1 - 8 1347
          AK150672.1 - 8 1268
          AK150984.1 - 8 1350
          AK150990.1 - 8 1350
          AK151006.1 - 8 1350
          AK151038.1 - 8 1248
          AK151093.1 - 8 1350
          AK151099.1 - 8 1350
          AK151105.1 - 8 1350
          AK151195.1 - 8 1248
          AK151369.1 - 8 1268
          AK151434.1 - 8 1268
          AK151521.1 - 8 1268
          AK151540.1 - 8 1238
          AK151563.1 - 8 1350
          AK151630.1 - 8 1248
          AK151679.1 - 8 1350
          AK151727.1 - 8 1350
          AK151772.1 - 8 1350
          AK151801.1 - 8 1238
          AK151828.1 - 8 1238
          AK151866.1 - 8 1350
          AK151870.1 - 8 1350
          AK151872.1 - 8 1350
          AK151893.1 - 8 1238
          AK152014.1 - 8 1350
          AK152035.1 - 8 1350
          AK152049.1 - 8 1350
          AK152053.1 - 8 1238
          AK152079.1 - 8 1350
          AK152238.1 - 8 1351
          AK152251.1 - 8 1348
          AK152350.1 - 8 1350
          AK152390.1 - 8 1189
          AK152481.1 - 8 1351
          AK152657.1 - 8 1350
          AK152706.1 - 8 1351
          AK152733.1 - 8 1248
          AK152775.1 - 8 1350
          AK152869.1 - 8 1350
          AK152956.1 - 8 1351
          AK153148.1 - 8 1350
          AK153242.1 - 8 1351
          AK153425.1 - 8 1350
          AK153428.1 - 8 1350
          AK159268.1 - 8 1350
          AK170486.1 - 8 1350
          BC003305.1 - 8 1338 3584788
          BC158040.1 - 8 1183 9088156
          CT010344.1 - 8 1151
          J03302.1 - 8 1127

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          lipase

          Gene Ontology Categories:

          Function(s) Process(es)

          lipid metabolic process
          fatty acid biosynthetic process
          positive regulation of macrophage derived foam cell differentiation
          positive regulation of cholesterol storage
          positive regulation of sequestering of triglyceride
          lipid catabolic process
          triglyceride biosynthetic process
          triglyceride catabolic process
          very-low-density lipoprotein particle remodeling
          triglyceride homeostasis
          lipoprotein lipase activity
          triglyceride lipase activity
          receptor binding
          protein binding
          heparin binding
          hydrolase activity
          triglyceride binding
          apolipoprotein binding
          carboxylic ester hydrolase activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-vnb2r:80/100.66.79.197:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0