Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lrrn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008516.4 | NP_032542.1 | 2 | 2058 | ||
GenBank mRNA | AK139355.1 | - | 2 | 2016 | ||
AK139965.1 | - | 2 | 2060 | |||
AK160841.1 | - | 2 | 1937 | |||
AK161885.1 | - | 1 | 560 | |||
AK173186.1 | - | 1 | 1303 | |||
BC031122.1 | - | 1 | 1333 | 4972803 | ||
BC058701.1 | - | 2 | 1991 | 6853964 | ||
D45913.1 | - | 2 | 2004 |