Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HSD17B1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000413.2 | NP_000404.2 | 5 | 1612 | ||
XM_005257292.3 | XP_005257349.1 | 4 | 1372 | |||
XM_006721857.3 | XP_006721920.1 | 4 | 1375 | |||
XM_006721858.3 | XP_006721921.1 | 4 | 1372 | |||
XM_006721859.3 | XP_006721922.1 | 4 | 1375 | |||
XM_011524729.1 | XP_011523031.1 | 5 | 1023 | |||
XM_011524730.1 | XP_011523032.1 | 5 | 1020 | |||
XM_011524731.1 | XP_011523033.1 | 5 | 1023 | |||
GenBank mRNA | AK127832.1 | - | 5 | 4364 | ||
AK300452.1 | - | 4 | 561 | |||
BC104752.1 | - | 5 | 670 | 8143755 | ||
BC111935.1 | - | 5 | 670 | 8327403 | ||
DQ469954.1 | - | 4 | 737 | |||
M36263.1 | - | 5 | 653 | |||
X13440.1 | - | 5 | 653 |