Search
Search

| Gene Symbol: Magea6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_020019.3 | NP_064403.1 | 2 | 821 | ||
| GenBank mRNA | BC109339.2 | - | 2 | 832 | 40052528 | |
| BC116902.1 | - | 1 | 731 | 40126221 | ||
| Gene Symbol: Magea1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_020015.2 | NP_064399.1 | 1 | 729 | ||
| GenBank mRNA | BC116904.1 | - | 1 | 729 | 40126223 | |
| Gene Symbol: Magea2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_020016.1 | NP_064400.1 | 4 | 966 | ||
| GenBank mRNA | BC106842.2 | - | 866 | 40045733 | ||
| Gene Symbol: Magea3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_020017.2 | NP_064401.2 | 1 | 731 | ||
| GenBank mRNA | AY438669.1 | - | 1 | 667 | ||
| BC119064.1 | - | 1 | 731 | 8733813 | ||
| BC119066.1 | - | 1 | 731 | 8733815 | ||