Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mitf | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001113198.1 | NP_001106669.1 | 10 | 1455 | ||
NM_001178049.1 | NP_001171520.1 | 10 | 1342 | |||
NM_008601.3 | NP_032627.1 | 9 | 1146 | |||
GenBank mRNA | AB221657.1 | - | 9 | 1008 | ||
AK154189.1 | - | 11 | 1457 | |||
AK155350.1 | - | 10 | 1456 | |||
BC108976.2 | - | 9 | 1110 | 40047440 | ||
CF733424.1 | - | 2 | 218 | |||
Z23066.1 | - | 9 | 1137 |