siRNA Details
Gene Symbol: Mknk2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AB164081.1 | - | 9 | 697 | ||
AK030830.1 | - | 4 | 249 | |||
AK154235.1 | - | 10 | 977 | |||
AK184143.1 | - | 3 | 134 | |||
AK194740.1 | - | 3 | 134 | |||
AK194893.1 | - | 3 | 134 | |||
BC010256.1 | - | 8 | 607 | 3257646 | ||
BY732164.1 | - | 9 | 700 | |||
Y11092.1 | - | 8 | 648 |
