Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other HSP90AB1 SIRNA Assays ›
          Gene Symbol:
          HSP90AB1 HSP90AB3P
          Gene Name:
          heat shock protein 90 alpha family class B member 1
          heat shock protein 90 alpha family class B member 3, pseudogene
          Gene Aliases:
          D6S182, HSP84, HSP90B, HSPC2, HSPCB
          HSP90BC, HSPCP1
          Chromosome Location:
          Chr.6: 44246166 - 44253888 on Build GRCh38
          Chromosome Location:
          Chr.4: 87891748 - 87894288 on Build GRCh38
          UniGene ID:
          Hs.509736
          Hs.744112
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          -
          siRNA ID s7000

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: HSP90AB1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001271969.1 NP_001258898.1 11 2191
          NM_001271970.1 NP_001258899.1 11 2119
          NM_001271971.1 NP_001258900.1 11 2034
          NM_001271972.1 NP_001258901.1 11 2148
          NM_007355.3 NP_031381.2 11 2178
          GenBank mRNA AF275719.1 - 9 1664
          AK025370.1 - 8 1481
          AK130494.1 - 3 628
          AK290734.1 - 6 1297
          AK294643.1 - 11 2005
          AK297367.1 - 11 1921
          AK312255.1 - 11 2039
          AL136543.1 - 11 2014
          AY359878.1 - 11 2052
          BC004928.1 - 11 2024 3621040
          BC009206.2 - 9 1718
          BC012807.2 - 11 2024 3530042
          BC014485.1 - 11 2019 4870198
          BC016753.2 - 11 2037 2987963
          BC068474.1 - 11 2042 4824686
          BX649079.1 - 11 2041
          M16660.1 - 11 2024
          Gene Symbol: HSP90AB3P
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          GenBank mRNA AY956764.1 - 4 1559

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          Hsp90 family chaperone

          Gene Ontology Categories:

          Function(s) Process(es)

          placenta development
          protein folding
          response to unfolded protein
          response to salt stress
          virion attachment to host cell
          regulation of protein ubiquitination
          positive regulation of protein binding
          negative regulation of proteasomal ubiquitin-dependent protein catabolic process
          positive regulation of phosphoprotein phosphatase activity
          positive regulation of protein import into nucleus, translocation
          Fc-gamma receptor signaling pathway involved in phagocytosis
          negative regulation of neuron apoptotic process
          positive regulation of nitric oxide biosynthetic process
          positive regulation of cell size
          protein stabilization
          regulation of interferon-gamma-mediated signaling pathway
          regulation of type I interferon-mediated signaling pathway
          cellular response to interleukin-4
          cellular response to organic cyclic compound
          positive regulation of protein serine/threonine kinase activity
          fibril organization
          cell-cell adhesion
          regulation of cellular response to heat
          UTP binding
          CTP binding
          double-stranded RNA binding
          protein binding
          ATP binding
          GTP binding
          sulfonylurea receptor binding
          protein kinase regulator activity
          kinase binding
          protein kinase binding
          MHC class II protein complex binding
          nitric-oxide synthase regulator activity
          TPR domain binding
          dATP binding
          histone deacetylase binding
          ion channel binding
          poly(A) RNA binding
          unfolded protein binding
          cadherin binding involved in cell-cell adhesion

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-vnb2r:80/100.66.79.197:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0