Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other NFE2L1 SIRNA Assays ›
          Gene Symbol:
          Nfe2l1
          Gene Name:
          nuclear factor, erythroid derived 2,-like 1
          Gene Aliases:
          AA408798, AW212678, LCR-F1, Lcrf1, NRF1, TCF-11, TCF11
          Chromosome Location:
          Chr.11: 96817413 - 96829968 on Build GRCm38
          UniGene ID:
          Mm.6743
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          siRNA ID s70519

          Purification
          Size
          Availability Made To Order
          Catalog # 4390771
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Nfe2l1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001130450.1 NP_001123922.1 3 1179
          NM_001130451.1 NP_001123923.1 2 113
          NM_001130452.1 NP_001123924.1 2 80
          NM_001130453.1 NP_001123925.1 2 130
          NM_001130454.1 NP_001123926.1 2 97
          NM_008686.3 NP_032712.2 3 1237
          GenBank mRNA AF071084.1 - 2 583
          AK131183.1 - 3 1172
          AK148471.1 - 2 97
          AK152040.1 - 3 1218
          AK153529.1 - 3 1219
          AK154167.1 - 3 1148
          AK194563.1 - 1 59
          BC043063.1 - 2 819 5705669
          BC047283.1 - 2 579 6489220
          BC099895.1 - 3 1203
          CF163519.1 - 2 125
          CJ144781.1 - 2 86
          CJ152359.1 - 2 101
          X78709.1 - 2 615

          Back To Top

          More Information


          Other Species Targeted

          This siRNA also targets the following transcripts in Human: NM_003204, AK302387, BC131769, AK090459, X77366, and others.
          This siRNA also targets the following transcripts in Rat: XM_006247238, XM_006247237, XM_006247239, XM_006247236, BC169095.

          Panther Classification:

          Molecular Function -

          basic leucine zipper transcription factor

          Gene Ontology Categories:

          Function(s) Process(es)

          regulation of transcription, DNA-templated
          transcription from RNA polymerase II promoter
          erythrocyte differentiation
          cellular response to oxidative stress
          positive regulation of transcription from RNA polymerase II promoter
          RNA polymerase II distal enhancer sequence-specific DNA binding
          transcriptional activator activity, RNA polymerase II transcription factor binding
          transcriptional activator activity, RNA polymerase II distal enhancer sequence-specific binding
          DNA binding
          transcription factor activity, sequence-specific DNA binding
          protein binding
          sequence-specific DNA binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-vnb2r:80/100.66.79.197:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0