Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other NGF SIRNA Assays ›
          Gene Symbol:
          Ngf
          Gene Name:
          nerve growth factor
          Gene Aliases:
          Ngfb
          Chromosome Location:
          Chr.3: 102469919 - 102521013 on Build GRCm38
          UniGene ID:
          Mm.1259
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          siRNA ID s70581

          Purification
          Size
          Availability Made To Order
          Catalog # 4390771
          Price 5,354.00
          Your Price
          Online offer:
          5354.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Ngf
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001112698.2 NP_001106168.1 3 675
          NM_013609.3 NP_038637.1 4 802
          GenBank mRNA AK144588.1 - 4 794
          BC011123.1 - 3 768
          K01759.1 - 3 769
          M14805.1 - 3 769
          M35075.1 - 4 790
          S62089.1 - 2 649
          V00836.1 - 4 790

          Back To Top

          More Information


          Other Species Targeted

          This siRNA also targets the following transcripts in Human: NM_002506, CR541855, X52599, BC032517, BT019733, and others.
          This siRNA also targets the following transcripts in Rat: NM_001277055, XM_006233052, XM_006233053, FQ232560.

          Panther Classification:

          Molecular Function -

          neurotrophic factor

          Gene Ontology Categories:

          Function(s) Process(es)

          transmembrane receptor protein tyrosine kinase signaling pathway
          cell-cell signaling
          peripheral nervous system development
          memory
          positive regulation of cell proliferation
          adult locomotory behavior
          extrinsic apoptotic signaling pathway via death domain receptors
          negative regulation of peptidase activity
          positive regulation of gene expression
          negative regulation of endopeptidase activity
          positive regulation of neuron projection development
          positive regulation of neuron maturation
          sensory perception of pain
          positive regulation of cell growth
          neuron projection development
          positive regulation of protein ubiquitination
          positive regulation of protein autophosphorylation
          intracellular signal transduction
          nerve growth factor signaling pathway
          negative regulation of neuron apoptotic process
          regulation of neuron differentiation
          positive regulation of neuron differentiation
          positive regulation of axon extension
          regulation of neurotransmitter secretion
          positive regulation of collateral sprouting
          neuron projection morphogenesis
          positive regulation of sequence-specific DNA binding transcription factor activity
          regulation of release of sequestered calcium ion into cytosol
          positive regulation of neurotrophin TRK receptor signaling pathway
          neuron apoptotic process
          cellular response to acid chemical
          extrinsic apoptotic signaling pathway in absence of ligand
          positive regulation of stem cell proliferation
          negative regulation of type B pancreatic cell apoptotic process
          enzyme inhibitor activity
          receptor signaling protein activity
          receptor binding
          nerve growth factor receptor binding
          growth factor activity
          metalloendopeptidase inhibitor activity
          peptidase inhibitor activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-green-55658555d6-qfdf2:80/100.66.76.55:80.
          git-commit: ec8e7df5f8fec8d765bd419205f1b5046a016d37
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.41.0-Offline