Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Enterprise Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other NOS2 SIRNA Assays ›
          Gene Symbol:
          Nos2
          Gene Name:
          nitric oxide synthase 2, inducible
          Gene Aliases:
          NOS-II, Nos-2, Nos2a, i-NOS, iNOS
          Chromosome Location:
          Chr.11: 78920859 - 78960232 on Build GRCm38
          UniGene ID:
          Mm.2893
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          siRNA ID s70693

          Purification
          Size
          Availability Made To Order
          Catalog # 4390771
          Price 5,354.00
          Your Price
          Online offer:
          5354.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Nos2
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_010927.3 NP_035057.1 14 1747
          GenBank mRNA AF065919.1 - 14 1744
          AF065920.1 - 14 1744
          AF065921.2 - 14 1744
          AF065922.2 - 13 1617
          AF065923.2 - 13 1617
          AY090567.1 - 13 1567
          BC062378.1 - 14 1756 4978648
          M84373.1 - 14 1748
          M87039.1 - 14 1819
          M92649.1 - 14 1755
          U43428.1 - 14 1635

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          oxidoreductase

          Gene Ontology Categories:

          Function(s) Process(es)

          ovulation from ovarian follicle
          response to hypoxia
          arginine catabolic process
          superoxide metabolic process
          nitric oxide biosynthetic process
          inflammatory response
          nitric oxide mediated signal transduction
          negative regulation of gene expression
          peptidyl-cysteine S-nitrosylation
          positive regulation of guanylate cyclase activity
          response to lipopolysaccharide
          cellular response to drug
          regulation of cell proliferation
          negative regulation of protein catabolic process
          defense response to bacterium
          negative regulation of blood pressure
          regulation of protein kinase activity
          positive regulation of vasodilation
          regulation of insulin secretion
          positive regulation of killing of cells of other organism
          oxidation-reduction process
          cellular response to lipopolysaccharide
          cellular response to interferon-gamma
          actin binding
          NADPH-hemoprotein reductase activity
          nitric-oxide synthase activity
          receptor binding
          iron ion binding
          protein binding
          calmodulin binding
          beta-catenin binding
          cAMP-dependent protein kinase regulator activity
          FMN binding
          oxidoreductase activity
          protein kinase binding
          heme binding
          tetrahydrobiopterin binding
          arginine binding
          protein homodimerization activity
          cadherin binding
          metal ion binding
          flavin adenine dinucleotide binding
          nitric-oxide synthase binding
          Hsp90 protein binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-ctdjb:80/100.66.76.55:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0