Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other NOS3 SIRNA Assays ›
          Gene Symbol:
          Nos3
          Gene Name:
          nitric oxide synthase 3, endothelial cell
          Gene Aliases:
          2310065A03Rik, Nos-3, eNOS, ecNOS
          Chromosome Location:
          Chr.5: 24364819 - 24384474 on Build GRCm38
          UniGene ID:
          Mm.258415
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          siRNA ID s70695

          Purification
          Size
          Availability Made To Order
          Catalog # 4390771
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Nos3
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_008713.4 NP_032739.3 8 1131
          GenBank mRNA AK077896.1 - 8 1133
          AK143221.1 - 7 903
          AK158106.1 - 8 1145
          BC052636.1 - 8 1118 6530765
          U53142.1 - 8 1119

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          oxidoreductase

          Gene Ontology Categories:

          Function(s) Process(es)

          angiogenesis
          ovulation from ovarian follicle
          in utero embryonic development
          blood vessel remodeling
          regulation of sodium ion transport
          regulation of the force of heart contraction by chemical signal
          regulation of systemic arterial blood pressure by endothelin
          arginine catabolic process
          nitric oxide biosynthetic process
          signal transduction
          nitric oxide mediated signal transduction
          negative regulation of cell proliferation
          negative regulation of muscle hyperplasia
          smooth muscle hyperplasia
          lung development
          positive regulation of guanylate cyclase activity
          lipopolysaccharide-mediated signaling pathway
          response to estradiol
          positive regulation of apoptotic process
          negative regulation of potassium ion transport
          endothelial cell migration
          positive regulation of angiogenesis
          negative regulation of blood pressure
          positive regulation of vasodilation
          negative regulation of smooth muscle cell proliferation
          regulation of blood vessel size
          negative regulation of hydrolase activity
          negative regulation of calcium ion transport
          oxidation-reduction process
          cellular response to mechanical stimulus
          cellular response to transforming growth factor beta stimulus
          actin binding
          actin monomer binding
          NADPH-hemoprotein reductase activity
          nitric-oxide synthase activity
          iron ion binding
          calmodulin binding
          beta-catenin binding
          FMN binding
          oxidoreductase activity
          heme binding
          tetrahydrobiopterin binding
          arginine binding
          cadherin binding
          metal ion binding
          flavin adenine dinucleotide binding
          NADP binding
          nitric-oxide synthase binding
          Hsp90 protein binding
          scaffold protein binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-hhcgv:80/100.66.79.225:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0