Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Enterprise Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other SLC11A2 SIRNA Assays ›
          Gene Symbol:
          Slc11a2
          Gene Name:
          solute carrier family 11 (proton-coupled divalent metal ion transporters), member 2
          Gene Aliases:
          DCT1, DMT1, Nramp2, mk, van
          Chromosome Location:
          Chr.15: 100387898 - 100424209 on Build GRCm38
          UniGene ID:
          Mm.234608
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          siRNA ID s70783

          Purification
          Size
          Availability Made To Order
          Catalog # 4390771
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Slc11a2
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001146161.1 NP_001139633.1 4 400
          NM_008732.2 NP_032758.2 4 400
          GenBank mRNA AF029758.1 - 4 335
          AK049856.1 - 4 348
          AK083478.1 - 4 346
          AK148276.1 - 4 348
          AK196267.1 - 2 241
          AK210047.1 - 3 271
          BC019137.1 - 4 374 5040976
          L33415.1 - 4 328

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          secondary carrier transporter

          Gene Ontology Categories:

          Function(s) Process(es)

          transition metal ion transport
          porphyrin-containing compound metabolic process
          porphyrin-containing compound biosynthetic process
          heme biosynthetic process
          transport
          ion transport
          cobalt ion transport
          copper ion transport
          iron ion transport
          manganese ion transport
          cellular copper ion homeostasis
          activation of cysteine-type endopeptidase activity involved in apoptotic process
          learning or memory
          response to manganese ion
          nickel cation transport
          vanadium ion transport
          ferrous iron transport
          lead ion transport
          proton transport
          cellular response to oxidative stress
          iron ion transmembrane transport
          copper ion transmembrane transport
          nickel cation transmembrane transport
          dendrite morphogenesis
          erythrocyte development
          iron ion homeostasis
          multicellular organismal iron ion homeostasis
          cadmium ion transmembrane transport
          ferrous iron import
          manganese ion transmembrane transport
          zinc II ion transmembrane transport
          cation transmembrane transport
          hydrogen ion transmembrane transport
          copper ion import into cell
          ferrous iron transmembrane transport
          transporter activity
          copper ion transmembrane transporter activity
          iron ion transmembrane transporter activity
          manganese ion transmembrane transporter activity
          iron ion binding
          copper ion binding
          zinc ion binding
          hydrogen ion transmembrane transporter activity
          cadmium ion transmembrane transporter activity
          cobalt ion transmembrane transporter activity
          ferrous iron transmembrane transporter activity
          lead ion transmembrane transporter activity
          nickel cation transmembrane transporter activity
          vanadium ion transmembrane transporter activity
          solute:proton symporter activity
          ferrous iron uptake transmembrane transporter activity
          nickel cation binding
          inorganic cation transmembrane transporter activity
          manganese ion binding
          cadmium ion binding
          transition metal ion transmembrane transporter activity
          cobalt ion binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-qvstf:80/100.66.73.196:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0