Search
Search

Ntn3| Gene Symbol: Ntn3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010947.3 | NP_035077.1 | 3 | 1556 | ||
| GenBank mRNA | AF128866.1 | - | 3 | 1396 | ||
| AF149094.1 | - | 3 | 1192 | |||
| AF152418.1 | - | 3 | 1311 | |||
| AK031199.1 | - | 3 | 1701 | |||
| AK083904.1 | - | 3 | 1289 | |||
| AK141856.1 | - | 3 | 1467 | |||
| BC053038.1 | - | 3 | 1648 | |||
| BC094362.1 | - | 3 | 1556 | 6830466 | ||