Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ogdh | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001252282.1 | NP_001239211.1 | 23 | 3062 | ||
NM_001252283.1 | NP_001239212.1 | 23 | 3050 | |||
NM_001252287.1 | NP_001239216.1 | 22 | 3093 | |||
NM_001252288.1 | NP_001239217.1 | 22 | 3065 | |||
NM_010956.4 | NP_035086.2 | 22 | 3017 | |||
GenBank mRNA | AK147289.1 | - | 22 | 3018 | ||
AK150009.1 | - | 22 | 2983 | |||
AK169286.1 | - | 22 | 3009 | |||
AK190496.1 | - | 3 | 340 | |||
AK220536.1 | - | 23 | 3024 | |||
BC012042.1 | - | 6 | 652 | |||
BC013670.1 | - | 22 | 2951 | 4188988 | ||
BC025040.1 | - | 7 | 817 | |||
BC029143.1 | - | 7 | 827 | |||
BC031165.1 | - | 7 | 796 | |||
BC049104.1 | - | 22 | 3018 | 6417392 | ||
BC057354.1 | - | 23 | 2978 | 6827509 | ||
BC061232.1 | - | 7 | 871 |