Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Olfr263 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010984.1 | NP_035114.1 | 1 | 420 | ||
GenBank mRNA | AF102516.1 | - | 1 | 231 | ||
BC116918.1 | - | 1 | 499 | 40126237 | ||
BC120698.1 | - | 1 | 499 | 40129621 |