Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PI4KA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_058004.3 | NP_477352.3 | 43 | 5151 | ||
XM_005261634.1 | XP_005261691.1 | 42 | 5058 | |||
XM_017028829.1 | XP_016884318.1 | 42 | 5085 | |||
XM_017028830.1 | XP_016884319.1 | 29 | 3307 | |||
GenBank mRNA | AB210002.1 | - | 43 | 5121 | ||
AF012872.1 | - | 42 | 4890 | |||
AK095811.1 | - | 3 | 241 | |||
AK124692.1 | - | 1 | 660 | |||
AK302424.1 | - | 43 | 5079 | |||
AK314781.1 | - | 15 | 1805 | |||
BC018120.1 | - | 13 | 1468 | |||
BC053654.1 | - | 12 | 1444 | |||
L36151.1 | - | 11 | 1437 |