Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other OPRM1 SIRNA Assays ›
          Gene Symbol:
          Oprm1
          Gene Name:
          opioid receptor, mu 1
          Gene Aliases:
          M-OR-1, MOP-R, MOR-1, MOR-1O, Oprm, mor, muOR
          Chromosome Location:
          Chr.10: 6758593 - 7038209 on Build GRCm38
          UniGene ID:
          Mm.438000
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          siRNA ID s71113

          Purification
          Size
          Availability Made To Order
          Catalog # 4390771
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Oprm1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001039652.2 NP_001034741.1 4 1488
          GenBank mRNA AF062753.1 - 4 1255
          AF074972.1 - 4 1090
          AF074974.1 - 5 1375
          AF167568.1 - 6 1561
          AK038389.1 - 4 1229
          AY160190.1 - 4 1255
          AY358080.1 - 4 974
          AY923181.1 - 3 902
          AY923182.1 - 3 1017
          BC119545.1 - 4 1255 40056952
          DQ363374.1 - 3 256
          DQ363375.1 - 3 537
          DQ868787.1 - 6 1776
          DQ868788.2 - 6 1671
          EF105311.1 - 5 1375
          EF105312.1 - 5 1375
          EF105313.1 - 6 1677
          EF105314.1 - 4 1255

          Back To Top

          More Information


          Other Species Targeted

          This siRNA also targets the following transcripts in Rat: NM_001038600, NM_001038601, AY225402, AY225403.

          Set Membership:

          G-protein coupled receptor

          Panther Classification:

          Molecular Function -

          G-protein coupled receptor transmembrane signal receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          signal transduction
          G-protein coupled receptor signaling pathway
          adenylate cyclase-activating dopamine receptor signaling pathway
          adenylate cyclase-inhibiting G-protein coupled receptor signaling pathway
          negative regulation of adenylate cyclase activity
          phospholipase C-activating G-protein coupled receptor signaling pathway
          positive regulation of cytosolic calcium ion concentration
          neuropeptide signaling pathway
          chemical synaptic transmission
          locomotory behavior
          sensory perception of pain
          negative regulation of cAMP biosynthetic process
          adenylate cyclase-inhibiting opioid receptor signaling pathway
          positive regulation of appetite
          cellular response to stress
          opioid receptor signaling pathway
          eating behavior
          positive regulation of cAMP-mediated signaling
          negative regulation of cAMP-mediated signaling
          negative regulation of nitric oxide biosynthetic process
          positive regulation of nitric oxide biosynthetic process
          response to ethanol
          behavioral response to ethanol
          positive regulation of neurogenesis
          negative regulation of cytosolic calcium ion concentration
          regulation of sensory perception of pain
          excitatory postsynaptic potential
          negative regulation of Wnt protein secretion
          positive regulation of ERK1 and ERK2 cascade
          calcium ion transmembrane transport
          cellular response to morphine
          regulation of N-methyl-D-aspartate selective glutamate receptor activity
          G-protein alpha-subunit binding
          signal transducer activity
          G-protein coupled receptor activity
          beta-endorphin receptor activity
          opioid receptor activity
          voltage-gated calcium channel activity
          protein binding
          protein C-terminus binding
          protein domain specific binding
          filamin binding
          G-protein beta-subunit binding
          morphine receptor activity
          neuropeptide binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-gwzxr:80/100.66.76.73:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0