Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other OTT SIRNA Assays ›
          Gene Symbol:
          Ott Gm10439 Gm15080 Gm15085 Gm15091 Gm15093 Gm15097 Gm15107 Gm15114 Gm15127 Luzp4
          Gene Name:
          ovary testis transcribed
          predicted gene 10439
          predicted gene 15080
          predicted gene 15085
          predicted gene 15091
          predicted gene 15093
          predicted gene 15097
          predicted gene 15107
          predicted gene 15114
          predicted gene 15127
          leucine zipper protein 4
          Gene Aliases:
          AV272316
          ENSMUSG00000072923
          OTTMUSG00000018964, Ott-like
          OTTMUSG00000018973
          OTTMUSG00000018981
          ENSMUSG00000072925, OTTMUSG00000018984
          OTTMUSG00000018988, Ott-like
          OTTMUSG00000019001, Ott-like
          OTTMUSG00000019010, Ott-like
          ENSMUSG00000072928, OTTMUSG00000019138
          ENSMUSG00000072927
          Chromosome Location:
          Chr.X: 149097952 - 149138152 on Build GRCm38
          Chromosome Location:
          Chr.X: 149594601 - 149636621 on Build GRCm38
          Chromosome Location:
          Chr.X: 147992993 - 148033174 on Build GRCm38
          Chromosome Location:
          Chr.X: 149447617 - 149487784 on Build GRCm38
          Chromosome Location:
          Chr.X: 149941387 - 149984982 on Build GRCm38
          Chromosome Location:
          Chr.X: 149272377 - 149305003 on Build GRCm38
          Chromosome Location:
          Chr.X: 149784424 - 149826694 on Build GRCm38
          Chromosome Location:
          Chr.X: 148180675 - 148224710 on Build GRCm38
          Chromosome Location:
          Chr.X: 148488949 - 148521446 on Build GRCm38
          Chromosome Location:
          Chr.X: 148663588 - 148703849 on Build GRCm38
          Chromosome Location:
          Chr.X: 148857694 - 148924139 on Build GRCm38
          UniGene ID:
          Mm.420007
          Mm.304331
          Mm.422381
          Mm.474426
          Mm.359506
          Mm.458016
          Mm.347476
          Mm.484933
          Mm.422946
          Mm.491025
          Mm.473681
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          -
          -
          -
          -
          -
          -
          -
          -
          -
          -
          siRNA ID s71161

          Purification
          Size
          Availability Made To Order
          Catalog # 4390771
          Price 5,354.00
          Your Price
          Online offer:
          5354.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Ott
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_011022.1 NP_035152.1 1287
          GenBank mRNA AK133020.1 - 2 158
          X96603.1 - 2 283
          X96604.1 - 933
          X96606.1 - 1287
          Gene Symbol: Gm10439
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001037716.2 NP_001032805.2 5 1208
          GenBank mRNA BC109132.1 - 5 1208 40038983
          Gene Symbol: Gm15080
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001198988.1 NP_001185917.1 5 1287
          Gene Symbol: Gm15085
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001122734.1 NP_001116206.1 1287
          Gene Symbol: Gm15091
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001122735.1 NP_001116207.1 5 1287
          GenBank mRNA BX513296.1 - 2 66
          Gene Symbol: Gm15093
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001099920.1 NP_001093390.1 1281
          Gene Symbol: Gm15097
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001198987.2 NP_001185916.2 5 1256
          Gene Symbol: Gm15107
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001081648.1 NP_001075117.1 1287
          GenBank mRNA BC109131.1 - 1256 40038981
          Gene Symbol: Gm15114
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001082966.1 NP_001076435.1 5 1233
          Gene Symbol: Gm15127
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001114400.2 NP_001107872.2 5 1281
          Gene Symbol: Luzp4
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001114383.1 NP_001107855.1 5 1287

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-ctdjb:80/100.66.76.55:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0