siRNA Details
Gene Symbol: Pcolce | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008788.2 | NP_032814.2 | 6 | 1008 | ||
GenBank mRNA | AB008548.1 | - | 6 | 932 | ||
AK142615.1 | - | 6 | 937 | |||
AK144392.1 | - | 6 | 1009 | |||
AK146829.1 | - | 6 | 1009 | |||
AK159430.1 | - | 6 | 996 | |||
AK159940.1 | - | 6 | 1000 | |||
AK162524.1 | - | 6 | 1012 | |||
CX210896.1 | - | 4 | 689 | |||
X57337.1 | - | 6 | 975 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Panther Classification:
