siRNA Details
Gene Symbol: Pdcd6ip | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164677.1 | NP_001158149.1 | 10 | 1408 | ||
NM_001164678.1 | NP_001158150.1 | 10 | 1393 | |||
NM_011052.2 | NP_035182.2 | 10 | 1393 | |||
GenBank mRNA | AF119955.1 | - | 10 | 1354 | ||
AJ005073.1 | - | 10 | 1241 | |||
AK029827.1 | - | 10 | 1360 | |||
AK031256.1 | - | 10 | 1394 | |||
AK033943.1 | - | 10 | 1382 | |||
AK129340.1 | - | 10 | 1363 | |||
AK145893.1 | - | 10 | 1381 | |||
AK147736.1 | - | 10 | 1366 | |||
AK167574.1 | - | 10 | 1351 | |||
AK169704.1 | - | 10 | 1394 | |||
BC026823.1 | - | 10 | 1335 | 4038684 | ||
BC051123.1 | - | 10 | 1244 | 6529850 |

More Information
Other Species Targeted

Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
