Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Peg3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008817.2 | NP_032843.2 | 9 | 1310 | ||
GenBank mRNA | AB003040.1 | - | 9 | 1307 | ||
AF038939.1 | - | 6 | 961 | |||
AK044602.1 | - | 10 | 1374 | |||
AK122238.1 | - | 9 | 1289 | |||
AK141217.1 | - | 9 | 1311 | |||
AK163845.1 | - | 9 | 1313 | |||
BC048778.1 | - | 4 | 687 | 6308481 | ||
BC072661.1 | - | 9 | 1298 | 30543809 | ||
BC085183.1 | - | 9 | 1295 | 30663888 | ||
BC089344.1 | - | 1 | 390 |