Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Phf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011078.3 | NP_035208.2 | 13 | 1882 | ||
GenBank mRNA | AB221590.1 | - | 2 | 268 | ||
AB221591.1 | - | 13 | 1766 | |||
AF043726.1 | - | 13 | 1882 | |||
AK172995.1 | - | 11 | 1589 | |||
BC051633.1 | - | 13 | 1880 | 6518591 |