Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Piga | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011081.2 | NP_035211.2 | 2 | 337 | ||
GenBank mRNA | AK032283.1 | - | 2 | 308 | ||
AK039802.1 | - | 2 | 337 | |||
AK161026.1 | - | 2 | 307 | |||
AK171453.1 | - | 2 | 307 | |||
BC138755.1 | - | 1 | 295 | 8861777 | ||
BC138758.1 | - | 1 | 295 | 8861780 | ||
BY270710.1 | - | 2 | 332 | |||
D26047.1 | - | 2 | 306 | |||
S78188.1 | - | 2 | 343 |