siRNA Details
Gene Symbol: Pou3f1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011141.2 | NP_035271.1 | 1 | 2074 | ||
GenBank mRNA | AK139418.1 | - | 1 | 813 | ||
AK140274.1 | - | 1 | 639 |

Gene Symbol: Pou3f1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011141.2 | NP_035271.1 | 1 | 2074 | ||
GenBank mRNA | AK139418.1 | - | 1 | 813 | ||
AK140274.1 | - | 1 | 639 |