siRNA Details
Gene Symbol: Pou3f3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008900.2 | NP_032926.2 | 1 | 1848 | ||
GenBank mRNA | BC079869.1 | - | 2101 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
