Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other ALDOA SIRNA Assays ›
          Gene Symbol:
          ALDOA
          Gene Name:
          aldolase, fructose-bisphosphate A
          Gene Aliases:
          ALDA, GSD12, HEL-S-87p
          Chromosome Location:
          Chr.16: 30053090 - 30070420 on Build GRCh38
          UniGene ID:
          Hs.513490
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID s72
          Validated

          Purification
          Size
          Availability Inventoried
          Catalog # 4390824
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: ALDOA
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_000034.3 NP_000025.1 14 2107
          NM_001127617.2 NP_001121089.1 9 1329
          NM_001243177.1 NP_001230106.1 10 1450
          NM_184041.2 NP_908930.1 9 1296
          NM_184043.2 NP_908932.1 9 1268
          XM_011545768.2 XP_011544070.1 11 1970
          GenBank mRNA AK026577.1 - 9 1145
          AK098778.1 - 12 2148
          AK301993.1 - 10 1262
          BC000367.2 - 6 708
          BC004333.1 - 14 2027 3630974
          BC010660.1 - 9 1214 3869603
          BC012880.1 - 9 1142 3459009
          BC013614.1 - 9 1225 3880857
          BC015888.2 - 9 1138 3911623
          BC016170.1 - 6 708
          BC016800.1 - 10 1166 4072487
          BU855133.1 - 4 415
          BX647566.1 - 9 1118
          CR536528.1 - 8 1020
          CR541880.1 - 8 1020
          FJ474908.1 - 10 1187
          M11560.1 - 10 1187
          M21190.1 - 5 606
          X05236.1 - 9 1166

          Back To Top

          More Information


          Matching Taqman Gene Expression Assay:


          Hs00605108_g1

          Panther Classification:

          Molecular Function -

          aldolase

          Gene Ontology Categories:

          Function(s) Process(es)

          platelet degranulation
          fructose metabolic process
          gluconeogenesis
          glycolytic process
          ATP biosynthetic process
          striated muscle contraction
          actin filament organization
          regulation of cell shape
          fructose 1,6-bisphosphate metabolic process
          muscle cell cellular homeostasis
          protein homotetramerization
          canonical glycolysis
          cell-cell adhesion
          actin binding
          fructose-bisphosphate aldolase activity
          protein binding
          cytoskeletal protein binding
          tubulin binding
          identical protein binding
          poly(A) RNA binding
          fructose binding
          cadherin binding involved in cell-cell adhesion

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-75crn:80/100.66.77.221:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0