siRNA Details
Gene Symbol: Pparg | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011146.3 | NP_035276.2 | 6 | 884 | ||
GenBank mRNA | AB644275.1 | - | 7 | 890 | ||
AY208183.1 | - | 6 | 884 | |||
AY208184.1 | - | 6 | 884 | |||
AY243585.1 | - | 6 | 884 | |||
BC021798.1 | - | 6 | 884 | 4191539 | ||
CT010340.1 | - | 6 | 839 | |||
EF062476.1 | - | 6 | 875 | |||
GQ868647.1 | - | 5 | 642 | |||
GQ868649.1 | - | 5 | 642 | |||
U01841.1 | - | 7 | 1239 | |||
U09138.1 | - | 6 | 878 | |||
U10374.1 | - | 5 | 749 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Panther Classification:
