Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ppox | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008911.2 | NP_032937.1 | 11, 12 | 1578 | ||
GenBank mRNA | AK134341.1 | - | 3107 | |||
AK142632.1 | - | 11, 12 | 1530 | |||
BC002047.1 | - | 11, 12 | 1533 | 3484079 | ||
D45185.1 | - | 11, 12 | 1304 | |||
U25114.1 | - | 11, 12 | 1553 |