Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pspn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008954.2 | NP_032980.2 | 2 | 201 | ||
GenBank mRNA | AF040960.1 | - | 2 | 198 | ||
BC130229.1 | - | 2 | 202 | 40141900 |