Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other PTGS2 SIRNA Assays ›
          Gene Symbol:
          Ptgs2
          Gene Name:
          prostaglandin-endoperoxide synthase 2
          Gene Aliases:
          COX2, Cox-2, PGHS-2, PHS-2, Pghs2, TIS10
          Chromosome Location:
          Chr.1: 150100124 - 150108017 on Build GRCm38
          UniGene ID:
          Mm.292547
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          siRNA ID s72392

          Purification
          Size
          Availability Made To Order
          Catalog # 4390771
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Ptgs2
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_011198.3 NP_035328.2 5, 6 745
          GenBank mRNA AK049923.1 - 5, 6 736
          AK144726.1 - 5, 6 770
          AK144956.1 - 5, 6 768
          AK149820.1 - 5, 6 770
          AK166221.1 - 5, 6 768
          AK172161.1 - 5, 6 770
          BC052900.1 - 5, 6 741 30059181
          M64291.1 - 5, 6 769
          M88242.1 - 5, 6 745
          M94967.1 - 5, 6 770

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          oxygenase

          Gene Ontology Categories:

          Function(s) Process(es)

          prostaglandin biosynthetic process
          angiogenesis
          lipid metabolic process
          fatty acid metabolic process
          fatty acid biosynthetic process
          prostaglandin metabolic process
          inflammatory response
          response to oxidative stress
          embryo implantation
          learning
          memory
          regulation of blood pressure
          positive regulation of cell proliferation
          negative regulation of cell proliferation
          positive regulation of vascular endothelial growth factor production
          positive regulation of cell death
          sensory perception of pain
          cyclooxygenase pathway
          keratinocyte differentiation
          bone mineralization
          ovulation
          positive regulation of fever generation
          positive regulation of synaptic plasticity
          negative regulation of synaptic transmission, dopaminergic
          response to lipopolysaccharide
          response to cytokine
          maintenance of blood-brain barrier
          regulation of cell proliferation
          positive regulation of NF-kappaB import into nucleus
          anagen
          positive regulation of apoptotic process
          positive regulation of nitric oxide biosynthetic process
          negative regulation of cell cycle
          positive regulation of vasoconstriction
          negative regulation of smooth muscle contraction
          positive regulation of smooth muscle contraction
          decidualization
          positive regulation of smooth muscle cell proliferation
          brown fat cell differentiation
          negative regulation of calcium ion transport
          positive regulation of synaptic transmission, glutamatergic
          oxidation-reduction process
          cellular response to fluid shear stress
          positive regulation of transforming growth factor beta production
          positive regulation of cell migration involved in sprouting angiogenesis
          positive regulation of fibroblast growth factor production
          positive regulation of brown fat cell differentiation
          positive regulation of platelet-derived growth factor production
          peroxidase activity
          prostaglandin-endoperoxide synthase activity
          lipid binding
          oxidoreductase activity
          oxidoreductase activity, acting on single donors with incorporation of molecular oxygen, incorporation of two atoms of oxygen
          enzyme binding
          heme binding
          protein homodimerization activity
          metal ion binding
          dioxygenase activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-st8h5:80/100.66.79.144:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0