Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pxn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011223.3 | NP_035353.1 | 8 | 1256 | ||
NM_133915.3 | NP_598676.2 | 9 | 1358 | |||
GenBank mRNA | AF293882.1 | - | 8 | 1181 | ||
AF293883.1 | - | 8 | 1178 | |||
AK148060.1 | - | 8 | 1242 | |||
AK149933.1 | - | 8 | 1221 | |||
AK167299.1 | - | 8 | 1195 | |||
AK171226.1 | - | 8 | 1244 | |||
AK171436.1 | - | 8 | 1221 | |||
BC003298.1 | - | 4 | 539 | |||
BC025493.1 | - | 8 | 1214 |